Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM3 SIRNA Assays ›
Gene Symbol:
Tpm3
Gene Name:
tropomyosin 3, gamma
Gene Aliases:
TM30nm, TMnm, Tm5NM, Tpm-5, Tpm5, Trop-5, gamma-TM, hTM30nm, hTMnm
Chromosome Location:
Chr.3: 90072651 - 90100902 on Build GRCm38
UniGene ID:
Mm.240839
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s81893
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tpm3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001253738.1
NP_001240667.1
3
543
NM_001253740.1
NP_001240669.1
3
543
NM_001271764.1
NP_001258693.1
3
543
NM_001293748.1
NP_001280677.2
4
526
NM_001293749.1
NP_001280678.1
3
543
NM_022314.3
NP_071709.3
4
526
GenBank mRNA
AF317223.1
-
4
440
AK050665.1
-
3
389
AK088111.1
-
3
403
AK088902.1
-
3
401
AK145238.1
-
3
400
AK145243.1
-
3
400
AK145804.1
-
3
400
AK146349.1
-
3
373
AK146557.1
-
3
525
AK151441.1
-
3
425
AK151500.1
-
3
400
AK151571.1
-
3
399
AK151578.1
-
3
400
AK151595.1
-
3
372
AK151602.1
-
3
400
AK151991.1
-
3
399
AK152260.1
-
3
399
AK152659.1
-
3
372
AK153031.1
-
3
372
AK160011.1
-
3
425
AK164086.1
-
3
400
AK165428.1
-
3
399
AK166836.1
-
3
387
AK167268.1
-
3
377
AK167534.1
-
3
405
AK167581.1
-
3
400
AK168843.1
-
3
368
AK169102.1
-
3
371
AK169245.1
-
3
376
AK169821.1
-
3
417
BC029186.1
-
4
483
4502880
BC055407.1
-
3669
BC092045.1
-
3
376
3591372
CX237327.1
-
3
365
U04541.1
-
4
496
X53753.1
-
3
411
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Process(es)
actin binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-8dfkb:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0