Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022017.3 | NP_071300.2 | 6 | 1207 | ||
GenBank mRNA | AB021875.2 | - | 5 | 1134 | ||
AF208026.1 | - | 6 | 1175 | |||
AF263522.1 | - | 5 | 1165 | |||
AF279672.1 | - | 6 | 1191 | |||
AJ296078.1 | - | 5 | 1126 | |||
AK083223.1 | - | 6 | 1178 | |||
AK158652.1 | - | 2 | 1060 | |||
BC127052.1 | - | 6 | 1172 | 40109678 |