Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdf2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022324.3 | NP_071719.1 | 1 | 240 | ||
GenBank mRNA | AB043006.1 | - | 1 | 180 | ||
BC023290.1 | - | 1 | 194 | |||
BC053425.1 | - | 1 | 222 | 6508268 | ||
BC053438.1 | - | 1 | 222 | |||
BY253823.1 | - | 1 | 241 |