Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lce1m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025420.2 | NP_079696.1 | 2 | 814 | ||
GenBank mRNA | AK003825.1 | - | 2 | 810 | ||
AK004318.1 | - | 2 | 816 | |||
AK019517.1 | - | 2 | 814 |