Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Metap1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025633.3 | NP_079909.1 | 6, 7 | 727 | ||
GenBank mRNA | AJ414378.1 | - | 3, 4 | 590 | ||
AK005241.1 | - | 6, 7 | 724 | |||
AK010067.1 | - | 6, 7 | 727 | |||
BC051534.1 | - | 3, 4 | 601 | 3466745 | ||
BC060739.1 | - | 5, 6 | 2235 |