Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025670.4 | NP_079946.2 | 5 | 655 | ||
GenBank mRNA | AK005157.1 | - | 5 | 641 | ||
AK017480.1 | - | 5 | 638 | |||
AK031705.1 | - | 5 | 657 | |||
AK035791.1 | - | 3 | 1950 | |||
AK051560.1 | - | 5 | 653 | |||
AK149939.1 | - | 5 | 651 | |||
AK150219.1 | - | 5 | 600 | |||
AK166141.1 | - | 5 | 648 | |||
BC004063.1 | - | 5 | 654 | 3590511 |