Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024181.2 | NP_077143.2 | 16 | 2066 | ||
GenBank mRNA | AF255459.1 | - | 16 | 1743 | ||
AF314002.1 | - | 15 | 1579 | |||
AK004617.1 | - | 16 | 1821 | |||
AK028184.1 | - | 16 | 1807 | |||
AK128945.1 | - | 16 | 1820 | |||
AK165652.1 | - | 16 | 1823 | |||
AK166502.1 | - | 16 | 1777 | |||
AK169020.1 | - | 16 | 1833 | |||
BC002207.1 | - | 7 | 628 | |||
BC033461.1 | - | 16 | 1805 | 4010341 |