Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013012.1 | NP_001013030.1 | 3 | 332 | ||
GenBank mRNA | AK077458.1 | - | 3 | 299 | ||
AK159396.1 | - | 2 | 157 | |||
BC038658.1 | - | 3 | 307 | |||
BC137753.1 | - | 3 | 254 | 8860772 |