Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dynlt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025975.5 | NP_080251.3 | 2 | 146 | ||
GenBank mRNA | AK010186.1 | - | 2 | 80 | ||
AK018842.1 | - | 2 | 124 | |||
AK088277.1 | - | 2 | 147 | |||
BC030032.1 | - | 2 | 73 | 1332100 |