Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp11l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026031.3 | NP_080307.1 | 2, 3 | 181 | ||
GenBank mRNA | AK012550.1 | - | 2, 3 | 182 | ||
AK145266.1 | - | 2, 3 | 214 | |||
AK147113.1 | - | 2, 3 | 212 | |||
BC043139.1 | - | 2, 3 | 161 | |||
BC061074.1 | - | 2, 3 | 210 | 6706808 |