Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026083.2 | NP_080359.2 | 19 | 5355 | ||
GenBank mRNA | AK016264.1 | - | 3 | 327 | ||
BC023707.1 | - | 6 | 1223 | |||
BC082576.1 | - | 8 | 2867 | |||
BC094575.1 | - | 12 | 3904 | |||
BC138264.1 | - | 19 | 5307 | 8861283 | ||
BG062845.2 | - | 3 | 371 |