Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fundc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026126.4 | NP_080402.3 | 4 | 297 | ||
GenBank mRNA | AK013460.1 | - | 4 | 275 | ||
AK014705.1 | - | 4 | 274 | |||
AK031214.1 | - | 4 | 274 | |||
AY234858.1 | - | 4 | 263 | |||
BC042794.1 | - | 3 | 254 | |||
BC049566.1 | - | 4 | 272 | 6742356 | ||
BC061464.1 | - | 4 | 267 | 6819539 | ||
BY124891.1 | - | 4 | 297 |