Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adck3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163290.1 | NP_001156762.1 | 13 | 1605 | ||
NM_023341.3 | NP_075830.2 | 13 | 1703 | |||
GenBank mRNA | AJ278735.1 | - | 13 | 1583 | ||
AK004177.1 | - | 6 | 588 | |||
AK004791.1 | - | 13 | 1645 | |||
AK014605.2 | - | 13 | 1645 | |||
AK186293.1 | - | 4 | 330 | |||
AK191635.1 | - | 4 | 328 | |||
AK192575.1 | - | 4 | 329 | |||
AK194890.1 | - | 4 | 328 | |||
AK196005.1 | - | 4 | 329 | |||
AK200253.1 | - | 4 | 329 | |||
AK202430.1 | - | 4 | 329 | |||
BC030937.1 | - | 13 | 1651 | 4922892 | ||
U31629.1 | - | 11 | 1244 |