Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290476.1 | NP_001277405.1 | 5 | 1234 | ||
NM_026167.4 | NP_080443.1 | 7 | 1253 | |||
GenBank mRNA | AK004677.1 | - | 7 | 1225 | ||
AK122491.1 | - | 5 | 1234 | |||
BC100393.1 | - | 6 | 1218 | 30737426 |