Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ero1lb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026184.2 | NP_080460.2 | 12 | 1236 | ||
GenBank mRNA | AF489856.1 | - | 12 | 1042 | ||
AK004981.1 | - | 1213 | ||||
AK006886.1 | - | 12 | 1096 | |||
AK011964.1 | - | 109 | ||||
AK041951.1 | - | 12 | 1212 | |||
AK160664.1 | - | 12 | 1252 | |||
BC058721.1 | - | 12 | 1236 | 5705507 | ||
BC111900.1 | - | 12 | 1206 | 40062137 | ||
BC112426.1 | - | 12 | 1206 | 40062136 | ||
BC145888.1 | - | 12 | 1187 | 40131191 | ||
BC145890.1 | - | 12 | 1187 | 40131193 |