Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MBD1 SIRNA Assays ›
Gene Symbol:
MBD1
Gene Name:
methyl-CpG binding domain protein 1
Gene Aliases:
CXXC3, PCM1, RFT
Chromosome Location:
Chr.18: 50266882 - 50281774 on Build GRCh38
UniGene ID:
Hs.405610
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s8547
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MBD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204136.1
NP_001191065.1
3
624
NM_001204137.1
NP_001191066.1
3
624
NM_001204138.1
NP_001191067.1
3
624
NM_001204139.1
NP_001191068.1
3
624
NM_001204140.1
NP_001191069.1
3
624
NM_001204141.1
NP_001191070.1
3
624
NM_001204142.1
NP_001191071.1
3
624
NM_001204143.1
NP_001191072.1
3
405
NM_001204151.2
NP_001191080.1
3
624
NM_001323942.1
NP_001310871.1
3
624
NM_001323947.1
NP_001310876.1
3
624
NM_001323949.1
NP_001310878.1
3
616
NM_001323950.1
NP_001310879.1
3
624
NM_001323951.1
NP_001310880.1
3
624
NM_001323952.1
NP_001310881.1
3
624
NM_001323953.1
NP_001310882.1
3
631
NM_001323954.1
NP_001310883.1
3
624
NM_002384.2
NP_002375.1
3
624
NM_015844.2
NP_056669.2
3
624
NM_015845.3
NP_056670.2
3
624
NM_015846.3
NP_056671.2
3
624
NM_015847.3
NP_056723.2
3
624
XM_005258271.2
XP_005258328.1
3
371
XM_006722456.2
XP_006722519.1
3
371
XM_011525991.1
XP_011524293.1
3
371
XM_011525993.2
XP_011524295.1
3
371
XM_011525994.2
XP_011524296.1
3
371
XM_011525998.2
XP_011524300.1
3
371
XM_011525999.1
XP_011524301.1
3
371
XM_011526001.1
XP_011524303.1
3
371
XM_011526002.1
XP_011524304.1
3
371
XM_011526003.2
XP_011524305.1
3
371
XM_011526006.1
XP_011524308.1
3
371
XM_011526007.1
XP_011524309.1
3
371
XM_017025751.1
XP_016881240.1
3
371
XM_017025752.1
XP_016881241.1
3
371
XM_017025753.1
XP_016881242.1
3
371
XM_017025754.1
XP_016881243.1
3
371
XM_017025755.1
XP_016881244.1
3
371
XM_017025756.1
XP_016881245.1
3
371
XM_017025757.1
XP_016881246.1
3
371
XM_017025758.1
XP_016881247.1
3
371
XM_017025759.1
XP_016881248.1
3
371
XM_017025760.1
XP_016881249.1
3
371
XM_017025761.1
XP_016881250.1
3
371
XM_017025762.1
XP_016881251.1
3
371
XM_017025763.1
XP_016881252.1
3
371
XM_017025764.1
XP_016881253.1
3
371
XM_017025765.1
XP_016881254.1
3
371
XM_017025766.1
XP_016881255.1
3
371
XM_017025767.1
XP_016881256.1
3
371
XM_017025768.1
XP_016881257.1
3
371
XM_017025769.1
XP_016881258.1
3
371
XM_017025770.1
XP_016881259.1
3
371
XM_017025771.1
XP_016881260.1
3
371
XM_017025772.1
XP_016881261.1
3
371
XM_017025773.1
XP_016881262.1
3
371
XM_017025774.1
XP_016881263.1
3
371
XM_017025775.1
XP_016881264.1
3
371
XM_017025776.1
XP_016881265.1
3
371
XM_017025777.1
XP_016881266.1
3
371
GenBank mRNA
AF078830.1
-
2
187
AF078831.1
-
2
187
AF078832.1
-
2
187
AF078833.1
-
2
187
AJ564845.1
-
3
326
AK225889.1
-
3
534
AK225948.1
-
3
330
AK291519.1
-
3
625
AK295401.1
-
3
356
AK300759.1
-
3
405
AK302004.1
-
3
316
BC033242.1
-
3
350
5432943
BC047353.1
-
3
309
EF488685.1
-
2
187
Y10746.1
-
3
326
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
methylation-dependent chromatin silencing
transcription from RNA polymerase II promoter
heart development
aging
response to fluoxetine
neuron differentiation
response to nutrient levels
response to estradiol
response to cocaine
regulation of DNA methylation
negative regulation of transcription, DNA-templated
negative regulation of astrocyte differentiation
DNA binding
chromatin binding
protein binding
zinc ion binding
methyl-CpG binding
double-stranded methylated DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f2ldd:80/10.239.165.14:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0