Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jagn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205025.1 | NP_001191954.1 | 2 | 515 | ||
NM_001243739.1 | NP_001230668.1 | 2 | 638 | |||
NM_026365.3 | NP_080641.1 | 2 | 631 | |||
GenBank mRNA | AK003673.1 | - | 2 | 557 | ||
AK017953.1 | - | 2 | 566 | |||
AK020149.1 | - | 2 | 587 | |||
AK141417.1 | - | 2 | 611 | |||
AK151252.1 | - | 2 | 593 | |||
AK170345.1 | - | 2 | 598 | |||
BC011296.1 | - | 2 | 598 | 3962988 | ||
BC025910.1 | - | 2 | 585 | 5252891 | ||
BC037019.1 | - | 2 | 585 | 5364053 | ||
CX236607.1 | - | 2 | 606 |