Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026498.2 | NP_080774.2 | 5 | 670 | ||
GenBank mRNA | AB049945.1 | - | 5 | 394 | ||
AK002993.1 | - | 5 | 507 | |||
AK152027.1 | - | 5 | 461 | |||
AK160207.1 | - | 5 | 450 | |||
AK166241.1 | - | 5 | 458 | |||
BC005701.1 | - | 5 | 412 | |||
BC028279.1 | - | 5 | 462 | 5370270 |