Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spire1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176832.2 | NP_789802.1 | 4 | 385 | ||
NM_194355.2 | NP_919336.1 | 4 | 483 | |||
GenBank mRNA | AK129296.1 | - | 6 | 990 | ||
BC058669.1 | - | 4 | 483 | |||
BC094375.1 | - | 4 | 539 | 30535423 | ||
BU058399.1 | - | 4 | 484 |