Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026598.3 | NP_080874.2 | 3 | 371 | ||
GenBank mRNA | AF243434.2 | - | 3 | 327 | ||
AK011237.1 | - | 3 | 330 | |||
AK019140.1 | - | 3 | 223 | |||
AK019963.1 | - | 3 | 366 | |||
AK044618.1 | - | 2 | 176 | |||
AK163371.1 | - | 3 | 330 | |||
BC027422.1 | - | 3 | 349 | 4035224 |