Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026660.2 | NP_080936.1 | 4 | 570 | ||
GenBank mRNA | AK018736.1 | - | 4 | 583 | ||
AK089055.1 | - | 4 | 587 | |||
AK152438.1 | - | 4 | 556 | |||
BC023012.1 | - | 4 | 570 | 5376230 | ||
BC064775.1 | - | 4 | 468 | 30023877 |