Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164742.1 | NP_001158214.1 | 3, 4 | 823 | ||
NM_026687.2 | NP_080963.2 | 4, 5 | 1128 | |||
GenBank mRNA | AB233454.1 | - | 4, 5 | 1128 | ||
AK002574.1 | - | 3, 4 | 1016 | |||
AK165409.1 | - | 3, 4 | 801 | |||
BC119526.1 | - | 3, 4 | 956 | 40056096 |