Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haghl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271433.1 | NP_001258362.1 | 2 | 297 | ||
NM_001271434.1 | NP_001258363.1 | 2 | 297 | |||
NM_001271435.1 | NP_001258364.1 | 2 | 297 | |||
NM_026897.3 | NP_081173.1 | 1 | 282 | |||
GenBank mRNA | AK005274.1 | - | 1 | 278 | ||
AK012748.1 | - | 2 | 294 | |||
AK034798.1 | - | 2 | 272 | |||
AK047517.1 | - | 2 | 252 | |||
AK079632.1 | - | 1 | 280 | |||
AK080454.1 | - | 1 | 512 | |||
BC005586.1 | - | 2 | 243 | |||
BC030466.1 | - | 2 | 282 | 5253213 | ||
BC083322.1 | - | 2 | 236 | 6820885 | ||
BY265614.1 | - | 2 | 298 | |||
BY293803.1 | - | 1 | 285 |