Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027150.1 | NP_081426.1 | 7 | 816 | ||
GenBank mRNA | AK009836.1 | - | 5 | 614 | ||
BC089626.1 | - | 7 | 786 | 6757304 | ||
BC131903.1 | - | 7 | 816 | 40130180 | ||
BC144730.1 | - | 6 | 846 | 9053278 |