Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133716.3 | NP_598477.2 | 8 | 1125 | ||
GenBank mRNA | BC031581.1 | - | 8 | 1114 | ||
BC032217.1 | - | 8 | 1101 | |||
BC052413.1 | - | 8 | 1125 | 5720177 |