Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028544.1 | NP_082820.1 | 9 | 2321 | ||
GenBank mRNA | AK011557.2 | - | 3 | 404 | ||
AK033276.1 | - | 7 | 1609 | |||
AK156602.1 | - | 9 | 2333 | |||
BC028444.1 | - | 393 | ||||
BC072584.1 | - | 6 | 1275 | |||
BC127059.1 | - | 7 | 1247 | 40109852 |