Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm13251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085522.2 | NP_001078991.1 | 4 | 2663 |
Gene Symbol: Znf41-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC096621.1 | - | 2718 | 30464103 |