Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004196.3 | NP_001004196.2 | 6 | 1099 | ||
NM_001318826.1 | NP_001305755.1 | 5 | 1019 | |||
NM_001318828.1 | NP_001305757.1 | 4 | 942 | |||
NM_001318830.1 | NP_001305759.1 | 4 | 937 | |||
NM_005944.6 | NP_005935.4 | 5 | 1024 | |||
XM_005247481.4 | XP_005247538.1 | 6 | 1273 | |||
XM_005247482.2 | XP_005247539.1 | 5 | 903 | |||
XM_005247485.2 | XP_005247542.1 | 5 | 1214 | |||
XM_005247486.2 | XP_005247543.1 | 4 | 711 | |||
XM_005247488.2 | XP_005247545.1 | 4 | 816 | |||
XM_017006452.1 | XP_016861941.1 | 6 | 1600 | |||
XM_017006453.1 | XP_016861942.1 | 4 | 866 | |||
XM_017006454.1 | XP_016861943.1 | 6 | 1589 | |||
XM_017006455.1 | XP_016861944.1 | 5 | 1224 | |||
XM_017006456.1 | XP_016861945.1 | 6 | 950 | |||
XM_017006457.1 | XP_016861946.1 | 5 | 898 | |||
XM_017006458.1 | XP_016861947.1 | 6 | 1590 | |||
XM_017006459.1 | XP_016861948.1 | 5 | 1209 | |||
GenBank mRNA | AF063591.1 | - | 5 | 815 | ||
AK074972.1 | - | 5 | 789 | |||
AK129797.1 | - | 4 | 706 | |||
AK293399.1 | - | 4 | 711 | |||
AK297194.1 | - | 6 | 890 | |||
AK312563.1 | - | 5 | 900 | |||
AL832529.1 | - | 5 | 809 | |||
AY603771.1 | - | 4 | 758 | |||
BC022522.1 | - | 5 | 817 | 4794050 | ||
BC031103.1 | - | 5 | 793 | 5301702 |