Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144877.1 | NP_659126.1 | 6 | 1656 | ||
GenBank mRNA | AK084750.1 | - | 6 | 1351 | ||
AK146751.1 | - | 1854 | ||||
AK168959.1 | - | 6 | 1863 | |||
AK169319.1 | - | 1855 | ||||
AK173044.1 | - | 1809 | ||||
BC014872.1 | - | 6 | 1656 | 3602469 |