Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023603.3 | NP_076092.1 | 9 | 2016 | ||
GenBank mRNA | AK018483.1 | - | 4 | 308 | ||
AK191209.1 | - | 2 | 98 | |||
AK192184.1 | - | 2 | 97 | |||
AK198254.1 | - | 2 | 98 | |||
AK211103.1 | - | 2 | 98 | |||
AK217233.1 | - | 3 | 178 | |||
AK219185.1 | - | 2 | 98 | |||
AY034062.1 | - | 9 | 1931 | |||
BC037486.1 | - | 7 | 729 | |||
BC049227.1 | - | 6 | 587 | |||
BC055468.1 | - | 5 | 374 | |||
BC080848.1 | - | 9 | 1982 | |||
BC089305.1 | - | 9 | 1982 | 30360144 | ||
CA319647.1 | - | 7 | 647 | |||
CJ304628.1 | - | 3 | 206 |