Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ulk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027895.1 | NP_082171.1 | 14 | 1444 | ||
GenBank mRNA | AK004783.1 | - | 12 | 2026 | ||
AK032030.1 | - | 12 | 1066 | |||
AK154642.1 | - | 14 | 1357 | |||
BC037093.1 | - | 13 | 1747 | |||
BC039958.1 | - | 10 | 806 | |||
BC151153.1 | - | 14 | 1370 | 9088047 |