Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2cd2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027909.2 | NP_082185.2 | 7 | 1356 | ||
GenBank mRNA | AK004920.1 | - | 7 | 1361 | ||
AK039220.1 | - | 7 | 1327 | |||
AK122237.1 | - | 6 | 1395 | |||
AK153804.1 | - | 7 | 1328 | |||
AK218954.1 | - | 1 | 30 | |||
BC035271.1 | - | 6 | 596 | |||
BC049875.1 | - | 3 | 283 | |||
BC056612.1 | - | 7 | 927 | 4952570 | ||
BC060156.1 | - | 7 | 1220 | 6810220 |