Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027994.1 | NP_082270.1 | 14 | 3803 | ||
GenBank mRNA | AK009683.1 | - | 3 | 442 | ||
AK078117.1 | - | 5 | 1030 | |||
AK129225.1 | - | 14 | 3764 | |||
BC027142.1 | - | 2 | 170 | |||
BC057457.1 | - | 7 | 2446 |