Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jmjd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028101.4 | NP_082377.2 | 4 | 355 | ||
GenBank mRNA | AK011308.1 | - | 4 | 352 | ||
AK148079.1 | - | 4 | 333 | |||
AK155810.1 | - | 4 | 357 | |||
AK172073.1 | - | 4 | 352 | |||
BC010800.1 | - | 4 | 317 | 4235072 |