Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173744.4 | NP_776105.1 | 2 | 273 | ||
GenBank mRNA | AK077810.1 | - | 2 | 187 | ||
AK166875.1 | - | 2 | 184 | |||
AK166972.1 | - | 2 | 180 | |||
BC099613.1 | - | 2 | 178 | 30031671 | ||
BC132465.1 | - | 2 | 186 | 40130742 | ||
BC132467.1 | - | 2 | 186 | 40130744 | ||
BC145286.1 | - | 2 | 210 | 9053840 |