Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080941.1 | NP_001074410.1 | 4 | 1010 | ||
GenBank mRNA | AK083314.1 | - | 5 | 1079 | ||
BC046484.1 | - | 4 | 994 |
Gene Symbol: Rsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013769.1 | NP_001013791.1 | 4 | 1010 | ||
GenBank mRNA | AK087838.1 | - | 1 | 634 | ||
BC103788.1 | - | 4 | 985 | 6333828 |