Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hepacam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175189.4 | NP_780398.2 | 3 | 687 | ||
GenBank mRNA | AK013637.1 | - | 3 | 688 | ||
AK081154.1 | - | 3 | 603 | |||
AK138438.1 | - | 2 | 211 | |||
AK158265.1 | - | 3 | 925 | |||
BC082537.1 | - | 3 | 601 | |||
BC139057.1 | - | 3 | 677 | 8862079 | ||
BC139058.1 | - | 3 | 677 | 8862080 |