Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028876.2 | NP_083152.1 | 2 | 424 | ||
GenBank mRNA | AK014065.1 | - | 2 | 367 | ||
AK014490.1 | - | 2 | 425 | |||
AK033463.1 | - | 2 | 352 | |||
AK078317.1 | - | 399 | ||||
AK140979.1 | - | 1 | 2503 | |||
AK151650.1 | - | 2 | 390 | |||
AK159813.1 | - | 2 | 382 | |||
AK170036.1 | - | 2 | 384 | |||
BC020076.1 | - | 2 | 386 | 3985782 | ||
BC132023.1 | - | 2 | 386 | 40130300 | ||
BC138226.1 | - | 2 | 386 | 8861245 |