Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Steap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103156.2 | NP_001096626.1 | 6 | 1660 | ||
NM_001103157.2 | NP_001096627.1 | 6 | 2015 | |||
NM_001285469.1 | NP_001272398.1 | 6 | 1569 | |||
NM_001285470.1 | NP_001272399.1 | 5 | 1534 | |||
NM_028734.5 | NP_083010.2 | 6 | 1717 | |||
GenBank mRNA | AK015015.1 | - | 3 | 687 | ||
AK052981.1 | - | 6 | 1719 | |||
AK086787.1 | - | 6 | 1688 | |||
AK145468.1 | - | 6 | 1607 | |||
AK162343.1 | - | 5 | 1496 | |||
AK165644.1 | - | 6 | 1980 | |||
AK166607.1 | - | 6 | 1606 | |||
BC150881.1 | - | 6 | 1576 | 9087792 | ||
BC150889.1 | - | 6 | 1576 | 9087800 | ||
BC150997.1 | - | 1627 | 9087910 | |||
BC151008.1 | - | 1627 | 9087921 |