Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028801.2 | NP_083077.2 | 11 | 1369 | ||
GenBank mRNA | AJ311906.1 | - | 1371 | |||
AK037730.1 | - | 11 | 1370 |