Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029078.3 | NP_083354.3 | 14 | 4691 | ||
GenBank mRNA | AK133347.1 | - | 10 | 3372 | ||
AK173035.1 | - | 14 | 4663 | |||
BC004648.1 | - | 7 | 1380 | |||
BC027361.1 | - | 7 | 2184 | |||
BC030492.1 | - | 5 | 641 |