Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Manf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029103.3 | NP_083379.2 | 4 | 509 | ||
GenBank mRNA | AB009291.1 | - | 4 | 483 | ||
AK014338.1 | - | 4 | 487 | |||
AK131997.1 | - | 4 | 510 | |||
AK165646.1 | - | 4 | 485 | |||
AK167580.1 | - | 4 | 509 | |||
BC022761.1 | - | 4 | 515 | |||
BC038901.1 | - | 4 | 497 | |||
BC048616.1 | - | 3 | 572 | 6742574 |