Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133858.4 | NP_598619.2 | 5 | 1592 | ||
NM_199475.1 | NP_955769.1 | 5 | 1592 | |||
GenBank mRNA | AB075603.1 | - | 4 | 603 | ||
AK036838.1 | - | 5 | 1584 | |||
AK087859.1 | - | 5 | 1405 | |||
AK169673.1 | - | 5 | 1592 | |||
AK208613.1 | - | 4 | 225 | |||
BC039762.1 | - | 4 | 410 | |||
BC051048.1 | - | 4 | 475 | |||
BC057901.1 | - | 3 | 121 | |||
BC093521.1 | - | 1 | 161 | 4007022 | ||
BC094388.1 | - | 5 | 1409 | 5708548 | ||
BC132301.1 | - | 4 | 837 | 40130578 | ||
BC144749.1 | - | 4 | 837 | 9053297 |