Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyrm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029327.3 | NP_083603.2 | 1, 2 | 48 | ||
GenBank mRNA | AK006294.1 | - | 1, 2 | 374 | ||
BC139403.1 | - | 1, 2 | 179 | 8862425 | ||
BC139406.1 | - | 1, 2 | 179 | 8862428 | ||
BY776521.1 | - | 1, 2 | 48 |