Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029478.3 | NP_083754.2 | 11 | 1161 | ||
GenBank mRNA | AB047550.1 | - | 11 | 1173 | ||
AB076609.1 | - | 10 | 1052 | |||
AK043091.1 | - | 11 | 1162 | |||
AK077443.1 | - | 11 | 1161 | |||
AK154616.1 | - | 1 | 411 | |||
AK159468.1 | - | 11 | 1176 | |||
AK167173.1 | - | 11 | 1169 | |||
AK167202.1 | - | 11 | 1169 | |||
AK211254.1 | - | 3 | 223 | |||
AX775963.1 | - | 11 | 1131 | |||
BC004013.1 | - | 11 | 1155 | 3493738 |