Gene Symbol: Vmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029478.3 | NP_083754.2 | 2 | 152 | ||
GenBank mRNA | AB047550.1 | - | 2 | 164 | ||
AK016312.1 | - | 2 | 169 | |||
AK043091.1 | - | 2 | 153 | |||
AK049559.1 | - | 2 | 145 | |||
AK050796.1 | - | 1 | 21 | |||
AK077443.1 | - | 2 | 152 | |||
AK159468.1 | - | 2 | 167 | |||
AK167173.1 | - | 2 | 161 | |||
AK167202.1 | - | 2 | 161 | |||
AK186168.1 | - | 1 | 72 | |||
AX775963.1 | - | 2 | 122 |