Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029553.3 | NP_083829.1 | 4 | 420 | ||
NM_198311.1 | NP_938053.1 | 5 | 412 | |||
GenBank mRNA | AK002597.1 | - | 4 | 406 | ||
AK032748.1 | - | 4 | 382 | |||
AK081697.1 | - | 4 | 387 | |||
BC017523.1 | - | 5 | 412 | 4527657 |