Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ip6k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029634.2 | NP_083910.2 | 2 | 457 | ||
GenBank mRNA | BC039922.1 | - | 2 | 468 | 2609803 |