Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135100.2 | NP_001128572.1 | 5 | 872 | ||
NM_029646.3 | NP_083922.1 | 5 | 872 | |||
GenBank mRNA | AK008082.1 | - | 5 | 820 | ||
BC016254.1 | - | 5 | 851 | 4912097 | ||
EU636995.1 | - | 4 | 286 | |||
EU636996.1 | - | 4 | 289 |