Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029711.1 | NP_083987.1 | 9 | 1089 | ||
GenBank mRNA | AK008777.1 | - | 5 | 535 | ||
AK083263.1 | - | 9 | 899 | |||
AK151513.1 | - | 9 | 1063 | |||
AK152356.1 | - | 10 | 914 | |||
AK170061.1 | - | 10 | 888 | |||
BC099504.1 | - | 8 | 741 | |||
BC115750.1 | - | 9 | 887 | 40051600 |